Learn More
CD138 Mouse anti-Human, PE, Clone: DL-101, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 550805
Description
Reacts with CD138 (Syndecan-1), an 85-92 kDa single chain transmembrane protein, expressed on multiple myeloma-derived cell lines and malignant plasma cell populations. It is also expressed on pre-B cells, immature B cells and plasma cells, but not on mature circulating B-lymphocytes. Syndecan-1 is a member of the family of transmembrane heparan sulfate proteoglycans. It is also expressed on some non-hematopoietic cells, including embryonic mesenchymal cells, vascular smooth muscle cells, endothelial and neural cells. CD138 binds to many extracellular matrix proteins through its heparan sulfate side-chains, like fibronectin, collagen types I, III and V, tenascin, thrombospondin, and antithrombin III. It is considered an extracellular matrix receptor that may serve as a co-receptor for fibroblast growth factor and related molecules. Monoclonal antibody DL-101 does not block the binding of clone B-B4 (another anti-syndecan-1 antibody) or Mi15 by flow cytometric analysis.
Specifications
| CD138 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| DL-101 | |
| Syndecan-1 | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.