missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD138; Clone: MI15; BD
Brand: BD Biosciences 563294
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone MI15
Isotype IgG1, κ
Species Reactivity Hu
Immunogen Human U266 and XG-1 Myeloma Cell Lines
Flow Cytometry
Specifications
| CD138 | |
| MI15 | |
| Human U266 and XG-1 Myeloma Cell Lines | |
| 50 Tests | |
| Primary | |
| IgG1 |
| Monoclonal | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction