missing translation for 'onlineSavingsMsg'
Learn More

CD138 Rat anti-Mouse, APC, Clone: 281-2, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 561705

 View more versions of this product

Product Code. 15801259

  • ONLINE EXCLUSIVE PRICE
    £152.00
    £145.83 / Each
    Save £6.17
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 281-2 antibody reacts with the core protein of CD138 (Syndecan-1), a cell-surface, integral membrane heparan sulfate- and chondroitin sulfate-containing proteoglycan that binds to interstitial extracellular matrix molecules. Syndecan-1 is predominantly expressed on epithelial cells, where its expression correlates with normal epithelial organization. It is also expressed on B lymphocytes at specific stages during their differentiation: precursor B cells in the bone marrow and antibody-secreting cells, including plasma cells, but not mature peripheral B cells. It is thus implicated in mediating B cell-matrix interactions. CD138 expression is also regulated during embryonic development, and the molecule shows a tissue- specific structural polymorphism resulting from different post-translational modifications. The 281-2 antibody may be used to detect the differently glycosylated forms, because it reacts with the core protein. Furthermore, the mAb detects the Syndecan-1 ectodomain which is cleaved from cell surfaces by a metalloproteinase.

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD138
Monoclonal
0.2mg/mL
SYND1; Syndecan-1; syn-1; Sdc1; Sstn; synstatin
Rat
Affinity Purified
RUO
Mouse
IgG2a κ
Flow Cytometry
281-2
APC
Aqueous buffered solution containing ≤0.09% sodium azide.
NAMRU mouse mammary gland epithelial cell line NMuMG
25 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.