Learn More
CD137 Ligand Mouse anti-Human, Unlabeled, Clone: C65-485, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559445
Description
Reacts with a type II membrane protein known as 4-1BB ligand. This molecule belongs to the TNF superfamily. It has been reported to have costimulatory function on activated T cells. Reports suggest that stimulation of activated T cells via 4-1BB ligand does not require the presence of other costimulatory molecules, including CD28. Some studies have reported human 4-1BB ligand to be expressed preferentially on primary B cells and B-cell lines. C65-485 antibody is able to react with recombinant human 4-1BB ligand (rh4-1BBL) bound to CD137 (4-1BB receptor) expressed on PHA-stimulated PBMC.
Specifications
| CD137 Ligand | |
| Monoclonal | |
| 0.5mg/mL | |
| 4-1BB Ligand | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| C65-485 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.