Learn More
CD137 Mouse anti-Human, Unlabeled, Clone: H41BB-M127, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 552532
Description
Human 4-1BB, also known as CDw137, is a type I transmembrane glycoprotein belonging to the tumor necrosis factor receptor (TNFR) superfamily. Naοve T cells lack the 4-1BB, which is not only induced upon T cell activation, but also remains on activated T cells. It has been reported that the expression of 4-1BB is restricted to primed CD4+ and CD8+ cells. When triggered by either 4-1BB ligand or antibody ligation, the 4-1BB molecule can act as a co-stimulator to induce T-cell activation and proliferation. In recent studies, 4-1BB has been emphasized for its effect on the development and survival of functionally active effector CD8 (+) T cells against tumors, virus infections, allogenic immune responses, and for potential therapeutic application.
Specifications
| CD137 | |
| Monoclonal | |
| 0.5mg/mL | |
| 4-1BB, CDw137 | |
| Mouse | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| ELISA | |
| H41BB-M127 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.