missing translation for 'onlineSavingsMsg'
Learn More

CD137 Mouse anti-Human, APC, Clone: 4B4-1, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 561702

 View more versions of this product

Product Code. 15871249

  • ONLINE EXCLUSIVE PRICE
    £73.85
    £70.52 / Each
    Save £3.33
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 4B4-1 monoclonal antibody specifically binds to CD137 which is also known as 4-1BB, and ILA (induced by lymphocyte activation). CD137 is a type I transmembrane glycoprotein that belongs to the TNF/NGF receptor family. It is encoded by TNFRSF9 (tumor necrosis factor receptor superfamily, member 9). CD137 is expressed on activated T cells, B cells, monocytes, and follicular dendritic cells. CD137 plays roles in the costimulation, differentiation and survival of T cells and B cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD137
Monoclonal
APC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Recombinant Human 4-1BB Fusion Protein
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
4B4-1
4-1BB
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.