Learn More
CD130 Mouse anti-Human, PE-CF594, Clone: AM64, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 564152
Description
The AM64 monoclonal antibody specifically binds to CD130 which is also known as gp130 (Membrane glycoprotein 130), IL6ST (Interleukin-6 signal transducer), and IL6RB/ IL-6Rβ (Interleukin-6 Receptor beta). CD130 is a 130 kDa type 1 membrane glycoprotein that belongs to the hemopoietic cytokine receptor superfamily. CD130 serves as a common signaling receptor for several cytokine receptor complexes including those that mediate the biological effects of IL-6, IL-11, IL-27, Cardiotrophin-1 (CT-1), Ciliary neurotrophic factor (CNTF), Leukemia inhibitory factor (LIF), and Oncostatin M (OSM). CD130 is widely expressed on a variety of cell types including T cells, activated B cells, monocytes, plasma cells, and endothelia. The AM64 antibody can be used in flow cytometry, immunoprecipitation, western blot, and immunohistochemistry on frozen sections.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).
Specifications
| CD130 | |
| Monoclonal | |
| PE-CF594 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Purified Human CD130 | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| AM64 | |
| IL6ST; IL6RB; IL-6Rβ; Interleukin-6 Receptor beta; gp130 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.