Learn More
CD13 Mouse anti-Human, PE-Cy7, Clone: WM15, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561599
Description
The WM15 monoclonal antibody specifically binds to the 150 kDa Type II integral membrane glycoprotein aminopeptidase N. This antibody binds to GM-progenitor cells, granulocytic and monocytic cells, and mast cells, but not to lymphocytes, platelets or erythrocytes. Aminopeptidase N is involved in the metabolism of many regulatory peptides.
Specifications
| CD13 | |
| Monoclonal | |
| PE-Cyanine7 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human AML Cells | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| WM15 | |
| ANPEP; APN; Aminopeptidase N; Alanyl aminopeptidase; LAP1; PEPN | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.