Learn More
CD127 Rat anti-Mouse, Brilliant Violet 421, Clone: SB/199, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 562959
Description
The SB/199 monoclonal antibody specifically binds to mouse CD127, the 65-75 kDa type-I transmembrane protein IL-7Rα. The high affinity IL-7 receptor complex is composed of at least two transmembrane proteins, IL-7Rα and CD132, the common γ chain. CD127 has some sequence homology to the cytokine receptor superfamily (also known as the hematopoietin receptor superfamily). Micelacking CD127 display profoundly impaired development of the B and T lymphoid cell lineages, but display no obvious non-lymphoid abnormalities. IL-7Rα is expressed on common lymphoid progenitors and early stages of B lineage development in the bone marrow, on the earliest thymocyte progenitors, on CD4-CD8- double-negative and CD4+ and CD8+ single-positive thymocytes, and on most peripheral T lymphocytes. Intestinal intraepithelial lymphocytes with low-density γδ TCR upregulate CD127 expression in response to IL-2, which may be secreted by neighboring αβ TCR-bearing T cells.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.
Specifications
| CD127 | |
| Monoclonal | |
| 0.2mg/mL | |
| Interleukin-7 receptor alpha chain; IL-7R alpha; IL-7RA; IL-7Rα; Il7r | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| SB/199 | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| BALB/c mouse pre-B cell line 1A9 | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.