Learn More
CD125 Mouse anti-Human, PE, Clone: A14, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555902
Description
Reacts with the α chain of the interleukin-5 receptor (IL-5R), a 55-60 kDa polypeptide, expressed on eosinophils and basophils. The IL-5R is composed of a specific α chain and a β chain which is homologous to the β chains of interleukin-3 (IL-3R) and GM-CSF receptors. IL-5 promotes the growth and differentiation of eosinophil precursors and activates mature eosinophils. Clone A14 was developed using IL-5R α -transfected cells as immunogen. This antibody binding can be blocked by IL-5.
Specifications
| CD125 | |
| Monoclonal | |
| 0.2mg/mL | |
| IL-5Rα | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| A14 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IL-5R α-transfected cells | |
| 0.2 mg | |
| Cell Biology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.