Learn More
CD123 Mouse anti-Human, PE, Clone: 9F5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 340545
Description
The CD123 antibody, clone 9F5, is derived from the hybridization of NS-1 mouse myeloma cells with spleen cells isolated from BALB/c mice immunized with IL-3Rα–transfected COS cells.
The CD123 antibody recognizes a 70-kilodalton (kDa) type I integral membrane glycoprotein, also known as the interleukin-3 receptor α-chain (IL-3Rα).
Specifications
| CD123 | |
| Monoclonal | |
| 12.5μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Human IL-3Rα Transfected Cell Line | |
| 50 Tests | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| 9F5 | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.