Learn More
CD121B Mouse anti-Human, Unlabeled, Clone: MNC2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 552401
Description
CD121b, also known as interleukin-1 receptor type II (IL-1RII), has a significant role in the inflammatory response. Interleukin-1 receptor type II acts as a decoy receptor to regulate prolonged persistence of IL-1 by binding to IL-1 and preventing the formation of signaling complexes. Membrane IL-1 type II receptor forms a 60 kDa soluble IL-1 receptor type II upon proteolytic cleavage from various cell types after activation with secondary, IL-1 induced cytokines. The soluble IL-1 type II receptor consists of 398 amino acids and 29 amino acids within a short cytoplasmic domain. The MNC2 monoclonal antibody reacts with recombinant and native soluble human IL-1 receptor type II.
Specifications
| CD121b | |
| Monoclonal | |
| 0.5mg/mL | |
| IL-1 Receptor Type II, IL-1 RII | |
| Mouse | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| ELISA | |
| MNC2 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.