missing translation for 'onlineSavingsMsg'
Learn More

CD120a Mouse anti-Human, Unlabeled, Clone: MABTNFR1-B1, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 550514

Product Code. 15836298

  • ONLINE EXCLUSIVE PRICE
    £374.00
    £287.56 / Each
    Save £86.44
    23% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The TNFR1-B1 antibody reacts with the extracellular domain of CD120A which is also known as the 55 kDa receptor for the human cytokines, tumor necrosis factor (TNF) and lymphotoxin-alpha (LT- α ). This receptor is referred to as the p55 or Type I Tumor Necrosis Factor Receptor (TNFRI). TNFRI are expressed by a variety of cell lines, tumor cells, and normal cell types including T cells, monocytes, macrophages, neutrophils, endothelial cells, hepatocytes, chondrocytes, and fibroblasts. Naive B cells express very low or undetectable levels of TNFRI whereas mature erythrocytes and platelets are uniformly negative for TNFRI expression. For the generation of the MABTNFR1-B1 hybridoma, BALB/c mice were immunized with purified, full-length human TNFRI protein that was expressed by insect cells that were infected with a recombinant human TNFRI-baculovirus expression vector. MABTNFR1-B1 specifically binds to natural and recombinant truncated forms of TNFRI.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD120a
Monoclonal
0.5mg/mL
TNF Receptor Type I
Mouse
0.5 mg
Immunology
Human
IgG2a κ
Flow Cytometry
MABTNFR1-B1
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.