missing translation for 'onlineSavingsMsg'
Learn More

CD11c Hamster anti-Mouse, PE, Clone: HL3, BD

Hamster Monoclonal Antibody

Brand:  BD Biosciences 561044

 View more versions of this product

Product Code. 15820719

  • ONLINE EXCLUSIVE PRICE
    £127.00
    £121.47 / Each
    Save £5.53
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The HL3 antibody reacts with the integrin αx chain of gp150, 95 (CD11c/CD18) which is expressed on dendritic cells and CD4- CD8+ intestinal intraepithelial lymphocytes (IEL) and is upregulated on IEL and lymph-node T cells following in vivo activation. CD11c is also found on human NK cells. Although its expression on mouse NK cells is not published, we have detected CD11c on mouse splenic NK cells. Cells of the monocyte/macrophage lineage have been reported to express low levels of CD11c. CD11c plays a role in binding of iC3b.

Host Species: Hamster
Clone: HL3
Isotype: IgG1, λ
Cross Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD11c
Monoclonal
0.2mg/mL
Cd11c; Itgax; Integrin alpha-X; Integrin αX; Cr4; Complement receptor 4
Hamster
Affinity Purified
RUO
Mouse
IgG1 λ2
Flow Cytometry
HL3
PE
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes
25 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.