Learn More
CD11a Mouse anti-Human, APC, Clone: HI111, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559875
Description
The HI111 monoclonal antibody specifically binds to CD11a, the 180 kDa integrin α chain. This type I transmembrane glycoprotein associates with CD18 (integrin β2) to form the heterodimeric glycoprotein CD11a/CD18. This heterodimer is also known as the lymphocyte (leukocytes) function associated antigen-1 (LFA-1) that is expressed on all leukocytes. LFA-1 is an adhesion molecule involved in lymphocyte and granulocyte functions. LFA-1 mediates adhesion of lymphoid cells to the vascular endothelium in association with its ligand, and the intracellular adhesion molecule-1 (ICAM-1), CD54. Other ligands are ICAM-2 (CD102) and ICAM-3 (CD50).
Specifications
| CD11a | |
| Monoclonal | |
| APC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| HI111 | |
| LFA-1α; Lymphocyte (Leukocyte) function-associated antigen 1 α chain; ITGAL | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Canine, Human, Rabbit | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.