Learn More
CD119 Mouse anti-Human, Unlabeled, Clone: GIR-94, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558935
Description
The GIR-94 antibody recognizes the extracellular region of the alpha chain subunit (80-95 kDa glycoprotein) of the human interferon-γ receptor (IFN-γRα; aka, CD119). The functionally active-form of the human IFN-γ receptor consists of two (or more) subunits, with IFN-γRα responsible for IFN-γβ chains required for the transduction of biologic responses. The IFN-γ receptor α chain (CD119) is expressed on the surface of most human cells (except mature erythrocytes) including monocytes, macrophages, T cells, B cells, NK cells, neutrophils, fibroblasts, epithelial cells, and endothelium. The ability of this antibody to bind to IFN-γ receptors of species other than human has not been determined. The immunogen used to generate this hybridoma was human IFN-γRαpurified from human placenta. The GIR-94 is a non-neutralizing antibody.
Specifications
| CD119 | |
| Monoclonal | |
| 0.5mg/mL | |
| IFN-γ Receptor α chain | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| GIR-94 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human IFN-γRα | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.