Learn More
CD117 Rat anti-Mouse, PE-Cy7, Clone: 2B8, BD
Rat Monoclonal Antibody
£89.45 - £215.63
Specifications
| Antigen | CD117 |
|---|---|
| Clone | 2B8 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15861229
|
BD Biosciences
561681 |
25 μg |
£93.10 £89.45 / Each Save £3.65 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15849238
|
BD Biosciences
558163 |
0.1 mg |
£225.00 £215.63 / Each Save £9.37 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 2B8 antibody reacts with CD117 (c-Kit), a transmembrane tyrosine-kinase receptor which is encoded by the Kit gene (formerly dominant white spotting, W). The c-Kit ligand (also known as steel factor, stem cell factor, and mast cell growth factor) encoded by the Kit1 gene (formerly steel, SI), is a co-mitogen for hematopoietic stem cells, myeloerythroid progenitors and a mast-cell differentiation factor. The KitW and Kit1SI mutant alleles have similar pleiotropic effects on the development of melanocytes, germ cells, and the hematopoietic system. In the adult bone marrow, CD117 is expressed on hematopoietic progenitor cells, including CD90 (Thy-1) low, TER-119-, CD45R/B220-, CD11b (Mac-1)-, Ly-6G (Gr-1)-, CD4-, CD8-, and Sca-1 (Ly-6A/E)+ multipotent hemotopoietic stem cells, progenitors committed to myeliod and/or erythroid lineages, and precursors of B and T lymphocytes. This widespread expression of CD117 in hematopoietic precursors is consistent with the participation of c-Kit and its ligand in the regulation of several hematopoietic lineages. Intrathymic expression of c-Kit and c-Kit ligand suggest that CD117 is also involved in the regulation of some events during the development of T lymphocytes. CD117 is also expressed by mast cells and by dendritic cells found in the periarteriolar lymphocytoc sheaths (T-cell areas) of splenic white pulp. The mAb 2B8 reportedly does not block the action of c-Kit. This clone 2B8 had been reported that there was cross-reactivity with rat.
Host Species: Rat
Clone: 2B8
Isotype: IgG2b κ
Species Reactivity: Mouse
Immunogen: Mouse Bone Marrow Mast Cells
Flow Cytometry
Specifications
| CD117 | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Mouse Bone Marrow Mast Cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 2B8 | |
| Flow Cytometry | |
| PE-Cyanine7 | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2b κ | |
| Affinity Purified | |
| c-KIT; W; SCFR; Stem Cell Factor Receptor; Sl; Steel Factor Receptor; Ssm |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.