Learn More
CD116 Mouse anti-Human, Brilliant Violet 650, Clone: hGMCSFR-M1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 564044
Description
The hGMCSFR-M1 antibody reacts with the subunit (GM-CSFR) of the human Granulocyte-Macrophage Colony-Stimulating Factor Receptor complex. This 75-85 kD subunit is also known as CD116. The hGMCSFR-M1 antibody was first clustered at the Fifth International Workshop on Human Leucocyte Differentiation Antigens. The GM-CSFR subunit associates with the 120-140 kD β c subunit (common subunit, CD131), that is shared with the receptors for interleukins IL-3 and IL-5. Both of the chains of the GM-CSFR complex are involved in ligand binding and intracellular signaling. The α chain appears to transmit most of the biological signals. CD116 is expressed by a variety of myeloid cell lines, hematopoietic and non-hematopoetic tumor cells, and normal cell types including monocytes, macrophages, neutrophils, eosinophils, myeloid dendritic cells, endothelial cells, fibroblasts, and placental trophoblasts. Lymphocytes are negative for GM-CSFR expression. Reports suggest that GM-CSFR plays a role in myeloid lineage growth and differentiation. The immunogen used to generate the hGMCSFR-M1 hybridoma was recombinant human GM-CSFR.
Specifications
| CD116 | |
| Monoclonal | |
| 0.2mg/mL | |
| CSF2RA; GM-CSF Receptor alpha; GM-CSFRα; GMCSFRA; GMR, SMDP4 | |
| Recombinant human GM-CSFR | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| hGMCSFR-M1 | |
| Brilliant Violet 650 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.