missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCAM-1 Mouse anti-Human, PE, Clone: STA, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516947
This item is not returnable.
View return policy
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
VCAM-1 is distributed across non-leukocyte and leukocyte cells. Its expression is reportedly correlated with adhesion of melanoma, Burkitt’s lymphoma, osteosarcoma, and kidney carcinoma but not colon carcinoma cells to endothelial cells which may determine the site of tumor metastasis.
Specifications
| VCAM-1 | |
| Monoclonal | |
| PE | |
| P19320 | |
| VCAM1 | |
| Cells of the DS6 T-cell line | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| STA | |
| VCAM1 | |
| CD106, DKFZp779G2333, INCAM-100, MGC99561, CD106 antigen, vascular cell adhesion protein 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 7412 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction