missing translation for 'onlineSavingsMsg'
Learn More

CD106 Rat anti-Mouse, Unlabeled, Clone: 429 (MVCAM.A), BD

Rat Monoclonal Antibody

£190.20 - £216.57

Specifications

Antigen CD106
Clone 429 (MVCAM.A)
Applications Flow Cytometry
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15897148
View Documents
BD Biosciences
553330
0.5 mg
ONLINE EXCLUSIVE PRICE
£225.00
£216.57 / Each
Save £8.43
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15846318
View Documents
BD Biosciences
550547
1 mL
ONLINE EXCLUSIVE PRICE
£198.00
£190.20 / Each
Save £7.80
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 429 monoclonal antibody specifically binds to both the long (∼110 kDa) transmembrane-spanning form and the truncated (∼47 kDa) GPI-linked form of vascular cell adhesion molecule-1 (VCAM-1, CD106). CD106 is constitutively expressed on bone marrow stromal cells, myeloid cells, and splenic dendritic cells. Its expression on endothelial cells is upregulated by inflammatory cytokines and in certain pathologic conditions. CD106 expression has also been detected on apoptotic thymocytes, splenocytes, and lymphoid cell lines. VCAM-1 is a counter-receptor for VLA-4 ( α; 4 β; 1 integrin) and LPAM-1 ( α; 4 β; 7 integrin), and the 429 antibody partially blocks VCAM-1-mediated binding functions. Source of the immunogen was the mouse preadipose cell line PA6.
Specifications

Specifications

CD106
Flow Cytometry
Unconjugated
RUO
Mouse
IgG2a κ
Affinity Purified
Vcam-1; Vascular cell adhesion molecule 1; Vascular cell adhesion protein 1
429 (MVCAM.A)
Monoclonal
Rat
Immunology
Mouse preadipose cell line PA6
Primary
Store undiluted at 4°C
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.