Learn More
Invitrogen™ VCAM-1 Monoclonal Antibody (1.4C3)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA112637
Product Code. 11518193
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Use on formalin-fixed sections requires boiling sections in 1 mM EDTA, pH 8.0 for 10-20 minutes followed by cooling at room temperature for 20 minutes. A suggested positive control for this product is tonsil tissue.
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VCAM-1
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
P19320
VCAM1
Stimulated HUVEC cells and mouse myeloma NS1 cells.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin)
1.4C3
Unconjugated
VCAM1
adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B
Mouse
Protein G
RUO
7412
-20°C, Avoid Freeze/Thaw Cycles