missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD105 Monoclonal Antibody (MEM-226), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119594
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.Specifications
CD105 | |
Monoclonal | |
FITC | |
ENG | |
AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
2022, 497010 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-226 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P17813 | |
ENG | |
Recombinant vaccinia virus containing the human CD105 cDNA. | |
100 Tests | |
Primary | |
Human, Rat | |
Antibody | |
IgG2a |