missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD105; Clone: 266; BD
Brand: BD Biosciences 563920
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: 266
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Human Umbilical Vein Endothelial Cells
Flow Cytometry (Intracellular Staining)
Specifications
| CD105 | |
| 266 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human |
| Monoclonal | |
| Brilliant Violet 421 | |
| Human Umbilical Vein Endothelial Cells | |
| 50 Tests | |
| Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction