Learn More
CD100 Monoclonal Antibody (A8), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516558
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. Once reconstituted, this product has a shelf life of approx. 18 months. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 (also known as semaphorin 4d), a 150 kDa homodimer, is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B cell signaling by augmenting B cell aggregation and survival and down regulating CD23 expression. CD72 is a lymphocyte receptor for the class IV semaphorin CD100, which is involved in regulating B cell signaling. CD100 is expressed on resting and PHA stimulated T cells. The protein is weakly expressed on NK cells, EBV transformed B cells, monocytes and tumor/peripheral blood T cell lines, and at higher density on activated T cells and B cells.
Specifications
| CD100 | |
| Monoclonal | |
| Conc. Not Determined | |
| PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
| Q92854 | |
| Sema4d | |
| Human PHA activated lymphocytes | |
| 100 Tests | |
| Primary | |
| Human, Non-human Primate | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| A8 | |
| APC | |
| Sema4d | |
| A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4D; semaphorin H; semaphorin-4D; Semaphorin-C-like 2; semaphorin-J; Semcl2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 10507 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.