Learn More
anti-CCT6B Matched Antibody Pair, Ab-1, Abnova
Brand: Abnova H00010693-AP11.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-CCT6B (100μg); Mouse monoclonal anti-CCT6B, IgG2aκ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This gene encodes a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
ELISA
Specifications
| CCT6B | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CCT6B. | |
| CCT-zeta-2/CCTZ-2/Cctz2/TCP-1-zeta-2/TSA303 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-CCT6B (100 μg) 2. Detection antibody: mouse monoclonal anti-CCT6B, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| CCT6B | |
| CCT6B | |
| Affinity Purified | |
| RUO | |
| 10693 | |
| Human | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.