missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CCR8, APC, Clone: 1055c, R&D Systems™
Rabbit Monoclonal Antibody
Specifications
| Antigen | CCR8 |
|---|---|
| Clone | 1055C |
| Immunogen | HEK293 human embryonic kidney cell line transfected with mouse CCR8 Accession # NP_031746 |
| Host Species | Rabbit |
| Primary or Secondary | Primary |
Description
CCR8 Monoclonal specifically detects CCR8 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CCR8 | |
| HEK293 human embryonic kidney cell line transfected with mouse CCR8 Accession # NP_031746 | |
| Primary | |
| Mouse | |
| APC | |
| 1237 | |
| Affinity Purified | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 1055C | |
| Rabbit | |
| Monoclonal | |
| IgG | |
| CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR8, CCR-8, CD198, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, ChemR1, CKRL1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCL1, GPRCY6, GPR-CY6, TER-1, TER1CC chemokine receptor CHEMR1 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Flow Cytometry | |
| RUO |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title