missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR2 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 34 publications
£207.00 - £507.00
Specifications
| Antigen | CCR2 |
|---|---|
| Clone | 48607 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18728317
![]() |
R&D Systems
FAB151P-025 |
25 Tests |
£207.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18735866
![]() |
R&D Systems
FAB151P-100 |
100 Tests |
£507.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR2 Monoclonal specifically detects CCR2 in Human samples. It is validated for Flow Cytometry.Specifications
| CCR2 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Human | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| CCR2 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Detects human CCR-2 transfectants but not the parental cell line or CCR-5 transfectants. |
| 48607 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| NP_001116868 | |
| 729230 | |
| NS0 mouse myeloma cell line transfected with human CCR2 Met1-Leu360 Accession # NP_001116868 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title