missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM30B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA524890
This item is not returnable.
View return policy
Description
Specifications
| TMEM30B | |
| Polyclonal | |
| Unconjugated | |
| Q3MIR4 | |
| TMEM30B | |
| KLH conjugated synthetic peptide between 42-71 amino acids from the N-terminal region of human CC50B | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TMEM30B | |
| CDC50B, cell cycle control protein 50B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 161291 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction