Learn More
Caveolin 2 Mouse, Unlabeled, Clone: 65, BD
Mouse Monoclonal Antibody
£151.41 - £448.00
Specifications
| Antigen | Caveolin 2 |
|---|---|
| Clone | 65 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15895579
|
BD Biosciences
610684 |
50 μg |
£158.00 £151.41 / 1 set Save £6.59 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15805589
|
BD Biosciences
610685 |
150 μg | |||||||
Description
Identified as a tyrosine phosphorylated protein in Rous sarcoma virus- transformed chick embryo fibroblasts (CEF), caveolin is now known to be ubiquitously expressed. Caveolin (also known as VIP21) localizes to non-clathrin membrane invaginations (caveolae) on the inner surface of the plasma membrane. This transmembrane protein plays a structural role in these specializations. Caveolin is also present at the trans-Golgi network (TGN) and similar quantities are found in apically and basolaterally destined transport vesicles. Caveolin is part of a complex containing glycosylphosphatidylinositol (GPI)-linked molecules and cytoplasmic signaling proteins. Caveolin is a transmembrane adaptor molecule that can simultaneously recognize GPI-linked proteins and interact with downstream cytoplasmic signaling molecules, such as c-yes, Annexin II, and hetero-trimeric G proteins. Although caveolin 2 is similar to caveolin 1 in distribution and tissue expression, caveolin 2 is most abundant in adipose tissue and its expression is up-regulated upon differentiation. This antibody has been reported to recognize an epitope located within region 79-88 of caveolin 2.
Specifications
| Caveolin 2 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 65 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.