Learn More
Caveolin 1 Mouse, Unlabeled, Clone: 2234, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610494
Additional Details : Weight : 0.01000kg
Description
Identified as a tyrosine phosphorylated protein in Rous sarcoma virus-transformed chick embryo fibroblasts (CEF), caveolin is now known to be ubiquitously expressed. Caveolin (also known as VIP21) localizes to non-clathrin membrane invaginations (caveolae) on the inner surface of the plasma membrane. This transmembrane protein plays a structural role in these specializations. Caveolin is also present at the trans-Golgi network (TGN) and similar quantities are found in apically and basolaterally destined transport vesicles. Caveolin is part of a complex containing glycosylphosphatidylinositol (GPI)-linked molecules and cytoplasmic signaling proteins. Caveolin is a transmembrane adaptor molecule that can simultaneously recognize GPI-linked proteins and interact with downstream cytoplasmic signaling molecules, such as c-yes, Annexin II, and hetero-trimeric G proteins. Caveolin-1 can generate two forms, α and β, due to alternate splicing of the mRNA. Caveolin-1 forms large lipid-binding homo-oligomers which are believed to play a role in caveolae formation. It may also function as a scaffolding protein which concentrates and organizes signaling molecules, a role supported by the fact that caveolin-1 interacts directly with inactive Ras and G-protein α subunits.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
Caveolin 1 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
150 μg | |
Cell Biology | |
Chicken, Human | |
IgG2a |
Western Blot | |
2234 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.