Learn More
Cathelicidin Monoclonal Antibody (H7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518048
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody is specific for human Cathelicidin (aa 115-121, NQARGSF). The immunogen was prepared using partially purified human neutrophil membrane proteins (heparin Ultrogellectin affinity purification fractions in octyl glucoside). This target has a predicted molecular weight of 19 kDa.
Cathelicidin Monoclonal antibody specifically detects Cathelicidin in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Cathelicidin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P49913 | |
CAMP | |
Partially purified human neutrophil membrane proteins (heparin Ultrogellectin affinity purification fractions in octyl glucoside) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
H7 | |
Unconjugated | |
CAMP | |
18 kDa cationic antimicrobial protein; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; Camp; CAP18; CAP-18; Cathelicidin antimicrobial peptide; cathelin-like protein; Cathelin-related antimicrobial peptide; CLP; Cnlp; CRAMP; FALL39; FALL-39; FALL-39 peptide antibiotic; hCAP18; hCAP-18; HSD26; LL37; MCLP | |
Mouse | |
Protein G | |
RUO | |
820 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |