Learn More
Invitrogen™ alpha Catenin Monoclonal Antibody (1G5)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA12000
Product Code. 11589292
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
MA1-2000 detects alpha catenin from human samples. MA1-2000 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~102 kDa protein representing alpha catenin from scc-1 cell extract. In Western blot, this antibody did not cross-react with mouse samples. MA1-2000 can also be used in immunoprecipitation and immunofluorescence procedures. The MA1-2000 immunogen is a 90 kDa MBP/alpha catenin fusion protein purified via an amylose column.
Alpha catenin is a member of the b-catenin family of cytosolic proteins. Alpha catenin interacts with N-cadherin and b-catenin on the cytosolic side of adherins junctions to mediate the interaction with the actin cytoskeleton. Alpha catenin is a homodimer in solution, but dissociates to form a 1:1 heterodimer with b-catenin in order to interact with cadherins. It also regulates morphological plasticity of synapses and cerebellar and hippocampal lamination during development. Alpha catenin also functions in the control of the startle modulation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha Catenin
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P26232, P35221
CTNNA1, CTNNA2
90 kDa MBP/alpha catenin fusion protein purified on amylose column.
100 μg
Primary
Human
Antibody
IgG1
Immunoprecipitation, Western Blot, Immunocytochemistry
1G5
Unconjugated
CTNNA2
102 kDa cadherin-associated protein; 2010010M04Rik; AA517462; AI481747; AI988031; alpha E catenin; alpha E-catenin; Alpha N-catenin; alpha(E)-catenin; alpha(N)-catenin; alpha-catenin; alpha-catenin-related protein; alpha-E-catenin; alphaE-catenin; alpha-N-catenin; alphaN-catenin; cadherin associated protein; Cadherin-associated protein; cadherin-associated protein, related; cancer/testis antigen 114; CAP102; CAPR; CAP-R; catenin; catenin (cadherin associated protein), alpha 1; catenin (cadherin associated protein), alpha 2; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin (cadherin-associated protein), alpha 2; catenin alpha 1; catenin alpha 2; Catenin alpha1; catenin alpha-1; Catenin alpha2; Catenin alpha-2; catenin, alpha 1; Catenin-alpha1; Catenin-alpha2; Catna; Catna1; Catna2; cdf; cerebellar deficient folia; chp; CT114; CTNNA1; Ctnna2; CTNR; FLJ36832; FLJ52416; HGNC:2509; MDPT2; Neural alpha-catenin; NY-REN-13 antigen; renal carcinoma antigen NY-REN-13
Mouse
Protein G
RUO
1495, 1496
-20°C, Avoid Freeze/Thaw Cycles