missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 9 Monoclonal Antibody (LAAP6 96 2-22), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA112562
Code : LD
Additional Details : Weight : 0.01000kg
Description
A suggested positive control for this product is MCF-7 cell lysate.
Caspase 9 Monoclonal antibody specifically detects Caspase 9 in Human samples. It is validated for Western Blot.Specifications
Caspase 9 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
P55211 | |
CASP9 | |
Recombinant human capase-9 protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
LAAP6 96 2-22 | |
Unconjugated | |
CASP9 | |
25 kDa caspase-9 dominant negative protein; AI115399; APAF3; APAF-3; apoptotic protease activating factor 3; apoptotic protease MCH-6; Apoptotic protease-activating factor 3; AW493809; cas9; CASP 9; CASP9; CASP-9; Casp9_v1; Casp-9-CTD; caspase 9; caspase 9, apoptosis-related cysteine peptidase; caspase 9, apoptosis-related cysteine protease; Caspase9; caspase-9; caspase-9 dominant negative form; Caspase-9 precursor; Caspase-9 subunit p10; Caspase-9 subunit p35; CASPASE-9c; caspase-9-carboxyl-terminal divergent; CTD; Ice-Lap6; ICE-like apoptotic protease 6; Mch6; OTTHUMP00000002324; PPP1R56; protein phosphatase 1, regulatory subunit 56; RNCASP9; RP11-265F14.3 | |
Mouse | |
Protein A/G | |
RUO | |
842 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |