Learn More
Caspase-3 Rabbit, Unlabeled, Clone: C92-605, BD
Rabbit Monoclonal Antibody
Brand: BD Biosciences 559565
Description
The caspase family of cysteine proteases plays a key role in apoptosis and inflammation. Caspase-3 is a key protease that is activated during the early stages of apoptosis and, like other members of the caspase family, is synthesized as an inactive pro-enzyme that is processed in cells undergoing apoptosis by self-proteolysis and/or cleavage by another protease. The processed forms of caspases consist of large (17-22 kDa) and small (10-12 kDa) subunits which associate to form an active enzyme. Active caspase-3, a marker for cells undergoing apoptosis, consists of a heterodimer of 17 and 12 kDa subunits which is derived from the 32 kDa pro-enzyme. Active caspase-3 proteolytically cleaves and activates other caspases, as well as relevant targets in the cytoplasm, e.g., D4-GDI and Bcl-2, and in the nucleus (e.g. PARP). This antibody has been reported to specifically recognize the active form of caspase-3 in human and mouse cells. It has not been reported to recognize the pro-enzyme form of caspase-3.
Specifications
| Caspase-3 | |
| Monoclonal | |
| 0.5mg/mL | |
| CPP32; Yama; Apopain | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| C92-605 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human Active Caspase-3 Fragment | |
| 25 μg | |
| Cell Biology | |
| Human, Mouse | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.