missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 1 p10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532144
Code : PN
Additional Details : Weight : 0.01000kg
Description
Predicted reactivity: Mouse (80%), Rat (83%), Pig (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Caspase-1 (CASP1) is a thiol protease that cleaves IL-1 beta between Asp and Ala. This cleavage causes the release of the mature cytokine which is involved in a variety of inflammatory processes. CASP1 is important for defense against pathogen. It cleaves and activates sterol regulatory element binding proteins (SREBPs). CASP1 also promotes apoptosis. Upon inflammasome activation, during DNA virus infection but not RNA virus challenge, CASP1 controls antiviral immunity through the cleavage of CGAS and renders it inactive.Specifications
Caspase 1 p10 | |
Polyclonal | |
Unconjugated | |
CASP1 | |
Casp1; CASP-1; CASP1 nirs variant 1; caspase 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1B converting enzyme; IL1BC; IL-1BC; IL1BCE; IL1B-convertase; interleukin 1 beta-converting enzyme; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1beta converting enzyme; interleukin-1 beta convertase; Interleukin-1 beta-converting enzyme; p45 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12362, 25166, 834 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P29452, P29466, P43527 | |
CASP1 | |
Recombinant fragment corresponding to a region within amino acids 268 and 404 of Human Caspase 1 p10 subunit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |