missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 1 p10 (Cleaved Ala317) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539882
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Caspases are a family of cysteine proteases that are key mediators of programmed cell death or apoptosis (1). The precursor form of all caspases is composed of a prodomain, and large and small catalytic subunits. The active forms of caspases are generated by several stimuli including ligand-receptor interactions, growth factor deprivation and inhibitors of cellular functions. All known caspases require cleavage adjacent to aspartates to liberate one large and one small subunit, which associate into a tetramer to form the active enzyme. Caspase-1/ICE (IL-1b converting enzyme) is similar to the cell death gene CED-3 of Caenorhabditilis elegans and regulates multiple proinflammatory cytokines, including interleukin-1b and interferon-gamma-inducing factor.Specifications
Caspase 1 p10 (Cleaved Ala317) | |
Polyclonal | |
Unconjugated | |
CASP1 | |
ICE, IL1BC, P45, CASP1 nirs variant 1, IL-1 beta-converting enzyme, IL1B-convertase, caspase 1, apoptosis-related cysteine peptidase interleukin 1, beta, convertase, caspase-1, interleukin 1, beta, convertase, interleukin 1-B converting enzyme | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
834 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P29466 | |
CASP1 | |
A synthetic peptide derived from human CASP1 | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |