Learn More
CARD11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529702
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Raji, K562, NCI-H929, BCL-1. Predicted reactivity: Mouse (96%), Chicken (87%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein has a domain structure similar to that of CARD14 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10.Specifications
CARD11 | |
Polyclonal | |
Unconjugated | |
CARD11 | |
0610008L17Rik; 2410011D02Rik; bcl10-interacting maguk protein 3; BENTA; BIMP3; Card11; CARD-containing MAGUK protein 1; Carma 1; CARMA1; caspase recruitment domain family member 11; caspase recruitment domain family, member 11; caspase recruitment domain-containing protein 11; IMD11; LOC100363332; MGC133069; PPBL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
108723, 84433 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.02 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8CIS0, Q9BXL7 | |
CARD11 | |
Recombinant fragment corresponding to a region within amino acids 25 and 275 of Human CARD11 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |