missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carboxypeptidase M Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521727
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, Rat liver. Predicted reactivity: Mouse (87%), Rat (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localized degradation of extracellular proteins.
Specifications
| Carboxypeptidase M | |
| Polyclonal | |
| Unconjugated | |
| CPM | |
| 1110060I01Rik; 5730456K23Rik; AA589379; Carboxypeptidase M; Cpm; E030045M14Rik; renal carboxypeptidase; urinary carboxypeptidase B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1368, 314855 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P14384 | |
| CPM | |
| Recombinant protein encompassing a sequence within the center region of human Carboxypeptidase M. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction