Learn More
CAPG (M01), Mouse anti-Human, Clone: 1F1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CAPG.
Brand: Abnova H00000822-M01.100ug
Description
This gene encodes a member of the gelsolin/villin family of actin-regulatory proteins. The encoded protein reversibly blocks the barbed ends of F-actin filaments in a Ca2+ and phosphoinositide-regulated manner, but does not sever preformed actin filaments. By capping the barbed ends of actin filaments, the encoded protein contributes to the control of actin-based motility in nonmuscle cells. Alternatively spliced transcript variants have been observed, but have not been fully described. [provided by RefSeq]
Sequence: MYTAIPQSGSPFPGSVQDPGLHVWRVEKLKPVPVAQENQGVFFSGDSYLVLHNGPEEVSHLHLWIGQQSSRDEQGACAVLAVHLNTLLGERPVQHREVQGNESDLFMSYFPRGLKYQEGGVESAFHKTSTGAPAAIKKLYQVKGKKNIRATERALNWDSFNTGDCFILDLGQNIFAWCGGKSNILERNKARDLALAIRDSERQGKAQVEIVTDGEEPAEMIQVLGPKPALKEGNPEEDLTADKANAQAAALYKVSDATGQMNLTKVADSSPFALELLISDDCFVLDNGLCGKIYIWKGRKANEKERQAALQVAEGFISRMQYAPNTQVEILPQGRESPIFKQFFKDWKSpecifications
| CAPG | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC000728 | |
| CAPG | |
| CAPG (AAH00728.1, 1 a.a. ∼ 348 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunofluorescence, Western Blot | |
| 1F1 | |
| Mouse monoclonal antibody raised against a full length recombinant CAPG. | |
| CAPG | |
| AFCP/MCP | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 822 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.