missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Canine IgM Secondary Antibody, FITC, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184637
This item is not returnable.
View return policy
Description
Goat anti Dog IgM polyclonal antibody recognizes canine IgM and shows no cross - reactivity with other canine immunoglobulin classes in immunoelectrophoresis and ELISA. Goat anti Dog IgM may cross react with IgM from other species.
Anti-Canine secondary antibodies are affinity-purified antibodies with well-characterized specificity for canine (dog) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Canine IgM | |
Polyclonal | |
FITC | |
Goat | |
Affinity chromatography | |
RUO | |
Canine | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen) | |
1 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide | |
Purified canine IgM. | |
1 mg | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG |