Learn More
calcium/calmodulin-dependent protein kinase kinase 2, beta, Mouse, Clone: 4C7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CAMKK2.
Brand: Abnova H00010645-M02.100ug
Description
The product of this gene belongs to the Serine/Threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. This protein plays a role in the calcium/calmodulin-dependent (CaM) kinase cascade by phosphorylating the downstream kinases CaMK1 and CaMK4. Seven transcript variants encoding six distinct isoforms have been identified for this gene. Additional splice variants have been described but their full-length nature has not been determined. The identified isoforms exhibit a distinct ability to undergo autophosphorylation and to phosphorylate the downstream kinases. [provided by RefSeq]
Sequence: MSSCVSSQPSSNRAAPQDELGGRGSSSSESQKPCEALRGLSSLSIHLGMESFIVVTECEPGCAVDLGLARDRPLEADGQEVPLDSSGSQARPHLSGRKLSLQERSQGGLAAGGSLDMNGRCICPSLPYSPSpecifications
| calcium/calmodulin-dependent protein kinase kinase 2, beta | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC026060 | |
| CAMKK2 | |
| CAMKK2 (AAH26060, 1 a.a. ∼ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4C7 | |
| Mouse monoclonal antibody raised against a partial recombinant CAMKK2. | |
| CAMKK2 | |
| CAMKK/CAMKKB/KIAA0787/MGC15254 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 10645 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.