Learn More
calcium/calmodulin-dependent protein kinase II alpha, Mouse, Clone: 2C12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CAMK2A.
Brand: Abnova H00000815-M03.100ug
Description
The product of this gene belongs to the serine/threonine protein kinases family, and to the Ca(2+)/calmodulin-dependent protein kinases subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. This calcium calmodulin-dependent protein kinase is composed of four different chains: alpha, beta, gamma, and delta. The alpha chain encoded by this gene is required for hippocampal long-term potentiation (LTP) and spatial learning. In addition to its calcium-calmodulin (CaM)-dependent activity, this protein can undergo autophosphorylation, resulting in CaM-independent activity. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: TTMLATRNFSGGKSGGNKKSDGVKESSESTNTTIEDEDTKVRKQEIIKVTEQLIEAISNGDFESYTKMCDPGMTAFEPEALGNLVEGLDFHRFYFENLWSRNSKPVSpecifications
| calcium/calmodulin-dependent protein kinase II alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC040457 | |
| CAMK2A | |
| CAMK2A (AAH40457, 305 a.a. ∼ 410 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 2C12 | |
| Mouse monoclonal antibody raised against a partial recombinant CAMK2A. | |
| CAMK2A | |
| CAMKA/KIAA0968 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 815 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.