missing translation for 'onlineSavingsMsg'
Learn More

CaM Kinase II Mouse, Unlabeled, Clone: 45, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611292

 View more versions of this product

Product Code. 15815869

  • ONLINE EXCLUSIVE PRICE
    £164.00
    £157.25 / 1 set
    Save £6.75
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ca2+;/calmodulin-dependent protein kinase II (CaM kinase II) is a multifunctional Ser/Thr kinase that regulates a number of cellular functions in response to increased intracellular Ca2+;. CaM kinase II is widely distributed, but is predominantly expressed in brain. It is involved in the regulation of neuronal functions such as neurotransmitter synthesis, neurotransmitter release, long-term potentiation, and formation of spatial learning. Neuronal CaM kinase II contains heteromers of two major subunits, α and β, at a ratio of 2:1 and homomers of α subunits. Each subunit has N-terminal ATP-binding and catalytic/regulatory domains and a C-terminal association domain. The regulatory domain consists of the autoinhibitory and calmodulin-binding sites. Assembly of the association domains of multiple subunits positions the regulatory domains for intersubunit autophosphorylation. After binding Ca2+;/calmodulin, CaM kinase II undergoes rapid autophosphorylation of the α and β subunits, which results in a substantial increase in its affinity for Ca2+;/calmodulin.

Host Species: Mouse
Clone: 45
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat CaM Kinase IIα aa. 448-460
Formula Weight [Chemical]: 52kDa

Immunofluorescence, Immunohistochemistry, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CaM Kinase II
Monoclonal
250μg/mL
Ca2+/calmodulin-dependent protein kinase II
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
In vitro Assay, Western Blot
45
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Rat CaM Kinase IIα aa. 448-460
50 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.