Learn More
CACNB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540992
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: EHSPLERDSL MPSDEASESS RQAWTGSSQR SSRHLEEDYA DAYQDLYQPH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
CACNB3 Polyclonal antibody specifically detects CACNB3 in Human samples. It is validated for Western Blot.Specifications
CACNB3 | |
Polyclonal | |
Unconjugated | |
Cacnb3 | |
Beta3; Ca(v)beta3; CAB3; CACH3B; Cacnb3; Cacnlb3; Calcium channel subunit beta 3; calcium channel voltage-dependent subunit beta 3; calcium channel, voltage-dependent, beta 3 subunit; calcium voltage-gated channel auxiliary subunit beta 3; cardiac calcium channel beta-subunit CaB3; Cav; CCHB3; voltage-dependent calcium channel beta3A subunit; voltage-dependent L-type calcium channel subunit beta-3 | |
Rabbit | |
Protein A | |
RUO | |
784 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P54284 | |
Cacnb3 | |
Synthetic peptide directed towards the C-terminal of human CACNB3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |