missing translation for 'onlineSavingsMsg'
Learn More

Cadherin-5 Mouse anti-Human, Unlabeled, Clone: 75, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610251

 View more versions of this product

Product Code. 15875359

  • ONLINE EXCLUSIVE PRICE
    £198.00
    £190.20 / 1 set
    Save £7.80
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cadherins are a family of transmembrane glycoproteins involved in the Ca2+- dependent cell-cell adhesion that occurs in many tissues. These proteins are similar in their domain structure (45-74% amino acid conservation), Ca2+ and protease-sensitivity, and molecular weight. Cadherin-5 (VE-Cadherin or CD144) is one of a number of cadherins (cadherin-4 through -11) whose cDNAs were isolated from rat brain and retina. These cadherins have a cytoplasmic domain that is highly conserved relative to previously identified cadherins, indicating that this domain is essential for cell adhesion activity. This function is mediated by cadherin interaction with cytoskeletal proteins. However, Cadherin-5's cytoplasmic domain has the lowest degree of homology with the other cadherins. Cadherin-5 is expressed in brain and various other tissues, including umbilical cord vein endothelial cells. A new type of adhering junction has been identified in certain vascular endothelial cells. These junctions are known as “complexus adherens” and are morphologically and compositionally distinct from desmosomes and zonula adherens junctions. The complexus adherens of endothelial cells lack desmosomal cadherins as well as E-Cadherin. However, these cells are rich in Cadherin-5 which colocalizes with desmoplakin and γ-Catenin (plakoglobin).

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cadherin-5
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Human
IgG1
Western Blot
75
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.