missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDH19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538856
This item is not returnable.
View return policy
Description
This gene is one of three related type II cadherin genes situated in a cluster on chromosome 18. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein containing five extracellular cadherin repeats. Loss of cadherins may be associated with cancer formation. Alternative splicing results in multiple transcript variants for this gene. [provided by RefSeq, Aug 2012].
Specifications
| CDH19 | |
| Polyclonal | |
| Unconjugated | |
| CDH19 | |
| CDH7L2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28513 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H159 | |
| CDH19 | |
| A synthetic peptide derived from the internal region of human CDH19 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction