missing translation for 'onlineSavingsMsg'
Learn More

anti-CACYBP, TBL1X Protein Protein Interaction Antibody Pair, Abnova™

Product Code. 16151514
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16151514 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16151514 Supplier Abnova Supplier No. DI0621.1Set
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

CACYBP rabbit purified polyclonal antibody (20μg); TBL1X mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols.

Host Species: Rabbit, Mouse
Cross Reactivity: Human

In situ PLA

Specifications

Antigen CACYBP
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene CACYBP
Gene Alias GIG5/MGC87971/PNAS-107/RP1-102G20.6/S100A6BP/SIP
Gene Symbols CACYBP
Host Species Mouse, Rabbit
Quantity 1 Set
Primary or Secondary Primary
Gene ID (Entrez) 27101/6907
Includes Antibody pair set content: 1. CACYBP rabbit purified polyclonal antibody (20 ug) 2. TBL1X mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type CACYBP & TBL1X Protein Protein Interaction Antibody Pair
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.