missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase VI Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526781
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VI Polyclonal Antibody for Western Blot, IHC (P), Flow
The protein encoded by this gene is one of several isozymes of carbonic anhydrase. This protein is found only in salivary glands and saliva and protein may play a role in the reversible hydratation of carbon dioxide though its function in saliva is unknown.
Specifications
| Carbonic Anhydrase VI | |
| Polyclonal | |
| Unconjugated | |
| CA6 | |
| CA6; CA-6; Car6; Carbonate dehydratase VI; carbonic anhydrase 6; Carbonic anhydrase VI; carbonic anhydrase VI nirs variant 2; Carbonic anhydrase6; CA-VI; DOC1; GUSTIN; salivary carbonic anhydrase; Secreted carbonic anhydrase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 765 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P23280 | |
| CA6 | |
| KLH conjugated synthetic peptide between 279-308 amino acids from the C-terminal region of human CA6. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction