Learn More
CA125, clone: M11, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512425
Description
CA 125 determinant is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.
MA5-12425 targets CA 125 in IHC (P) applications and shows reactivity with Human samples.The MA5-12425 immunogen is cA 125 antigen derived from ascites fluid.
Specifications
| CA125 | |
| Monoclonal | |
| Unconjugated | |
| MUC16 | |
| CA-125, CA125 ovarian cancer antigen, MUC-16, mucin-16, ovarian cancer-related tumor marker CA125, ovarian carcinoma antigen CA125 | |
| Mouse | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| M11 | |
| tissue culture supernatant with 0.09% sodium azide | |
| Q8WXI7 | |
| MUC16 | |
| CA 125 antigen derived from ascites fluid | |
| RUO | |
| 94025 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.