missing translation for 'onlineSavingsMsg'
Learn More
Learn More
chromosome 9 open reading frame 68, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human C9orf68 protein.
Brand: Abnova H00055064-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MPLEVVVELQIRAISCPGVFLPGKQDVYLGVYLMNQYLETNSFPSAFPIMIQESMRFEKVFESAVDPGAVVDLLEMWDELAYYEENTRDFLFPEPKLTPSHPRRCREVLMKTALGFPGIAPKIEFSTRTAIRECVFLHRNRFLEERHESRRPLSTSHEPIFPLNTIKMKLRENNLNRLPKGMQARAPSQYSTRHFFQDQPAQLNLGNNFKISGGSKPPFVVRHVDSAKPFGENISEHHLRRSGRKSKFSDFPFPTRRASSLDSLAANVKVIKEPDERIVLRSDSSSCLDSSQFGKSSSSKQGDADFHGKASFATYQHSTSPGPLDQPLLRERFHPGSQSTWKNIHERVCSLLTSHRAQLHQNKEDSTSEVNYIIERPSYPLKKYSLHEQRYFSpecifications
| chromosome 9 open reading frame 68 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human C9orf68 protein. | |
| C9orf68 | |
| FLJ10058/bA6J24.2 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 55064 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC034293 | |
| C9orf68 | |
| C9orf68 (AAH34293, 1 a.a. ∼ 392 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction